Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS1 SIRNA Assays ›
Gene Symbol:
HPS1
Gene Name:
HPS1, biogenesis of lysosomal organelles complex 3 subunit 1
Gene Aliases:
BLOC3S1, HPS
Chromosome Location:
Chr.10: 98415068 - 98446963 on Build GRCh38
UniGene ID:
Hs.404568
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS142542
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000195.4
NP_000186.2
4
429
NM_001311345.1
NP_001298274.1
4
438
NM_001322476.1
NP_001309405.1
4
358
NM_001322477.1
NP_001309406.1
4
438
NM_001322478.1
NP_001309407.1
4
358
NM_001322479.1
NP_001309408.1
4
438
NM_001322480.1
NP_001309409.1
4
358
NM_001322481.1
NP_001309410.1
4
438
NM_001322482.1
NP_001309411.1
4
438
NM_001322483.1
NP_001309412.1
4
358
NM_001322484.1
NP_001309413.1
4
438
NM_001322485.1
NP_001309414.1
4
429
NM_001322487.1
NP_001309416.1
4
438
NM_001322489.1
NP_001309418.1
4
438
NM_001322490.1
NP_001309419.1
4
438
NM_001322491.1
NP_001309420.1
4
358
NM_001322492.1
NP_001309421.1
4
358
NM_182639.3
NP_872577.1
4
438
XM_005269757.4
XP_005269814.1
4
371
XM_017016170.1
XP_016871659.1
4
345
XM_017016171.1
XP_016871660.1
4
434
XM_017016172.1
XP_016871661.1
4
406
XM_017016173.1
XP_016871662.1
4
436
GenBank mRNA
AK295821.1
-
4
394
AL702912.1
-
4
423
BC000175.2
-
4
388
BI518436.1
-
4
348
DA385131.1
-
4
436
DA479757.1
-
4
358
KU178114.1
-
2
167
KU178115.1
-
2
167
KU178116.1
-
2
167
KU178117.1
-
2
167
KU178118.1
-
2
167
KU178119.1
-
2
167
KU178120.1
-
2
167
KU178121.1
-
2
167
KU178122.1
-
2
167
U65676.1
-
4
373
U96721.1
-
4
401
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lysosome organization
visual perception
response to stimulus
protein binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0