Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFNA16 SIRNA Assays ›
Gene Symbol:
IFNA16
Gene Name:
interferon alpha 16
Gene Aliases:
IFN-alpha-16, IFN-alphaO
Chromosome Location:
Chr.9: 21216373 - 21217311 on Build GRCh38
UniGene ID:
Hs.56303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS142615
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFNA16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002173.3
NP_002164.1
1
235
GenBank mRNA
M28585.1
-
1
235
X00140.1
-
1
235
Back To Top
More Information
Panther Classification:
Molecular Function -
interferon superfamily
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
blood coagulation
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
innate immune response
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
cytokine activity
type I interferon receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0