Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114396.1 | NP_001107868.1 | 7 | 723 | ||
NM_002262.3 | NP_002253.1 | 6 | 786 | |||
NM_007334.2 | NP_031360.1 | 5 | 693 | |||
XM_017019291.1 | XP_016874780.1 | 7 | 710 | |||
GenBank mRNA | AB009597.1 | - | 5 | 470 | ||
AF498040.1 | - | 6 | 604 | |||
AF498041.1 | - | 5 | 505 | |||
AJ000001.1 | - | 6 | 557 | |||
AK291506.1 | - | 7 | 728 | |||
AY227806.1 | - | 7 | 723 | |||
AY327501.1 | - | 5 | 591 | |||
BC028009.1 | - | 6 | 599 | 5213081 | ||
U30610.1 | - | 6 | 610 |