Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBD1 SIRNA Assays ›
Gene Symbol:
MBD1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
CXXC3, PCM1, RFT
Chromosome Location:
Chr.18: 50266882 - 50281774 on Build GRCh38
UniGene ID:
Hs.405610
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS142880
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204136.1
NP_001191065.1
2
491
NM_001204137.1
NP_001191066.1
2
491
NM_001204138.1
NP_001191067.1
2
491
NM_001204139.1
NP_001191068.1
2
491
NM_001204140.1
NP_001191069.1
2
491
NM_001204141.1
NP_001191070.1
2
491
NM_001204142.1
NP_001191071.1
2
491
NM_001204143.1
NP_001191072.1
2
272
NM_001204151.2
NP_001191080.1
2
491
NM_001323942.1
NP_001310871.1
2
491
NM_001323947.1
NP_001310876.1
2
491
NM_001323949.1
NP_001310878.1
2
491
NM_001323950.1
NP_001310879.1
2
491
NM_001323951.1
NP_001310880.1
2
491
NM_001323952.1
NP_001310881.1
2
491
NM_001323953.1
NP_001310882.1
2
491
NM_001323954.1
NP_001310883.1
2
491
NM_002384.2
NP_002375.1
2
491
NM_015844.2
NP_056669.2
2
491
NM_015845.3
NP_056670.2
2
491
NM_015846.3
NP_056671.2
2
491
NM_015847.3
NP_056723.2
2
491
XM_005258271.2
XP_005258328.1
2
238
XM_006722456.2
XP_006722519.1
2
238
XM_011525991.1
XP_011524293.1
2
238
XM_011525993.2
XP_011524295.1
2
238
XM_011525994.2
XP_011524296.1
2
238
XM_011525998.2
XP_011524300.1
2
238
XM_011525999.1
XP_011524301.1
2
238
XM_011526001.1
XP_011524303.1
2
238
XM_011526002.1
XP_011524304.1
2
238
XM_011526003.2
XP_011524305.1
2
238
XM_011526006.1
XP_011524308.1
2
238
XM_011526007.1
XP_011524309.1
2
238
XM_017025751.1
XP_016881240.1
2
238
XM_017025752.1
XP_016881241.1
2
238
XM_017025753.1
XP_016881242.1
2
238
XM_017025754.1
XP_016881243.1
2
238
XM_017025755.1
XP_016881244.1
2
238
XM_017025756.1
XP_016881245.1
2
238
XM_017025757.1
XP_016881246.1
2
238
XM_017025758.1
XP_016881247.1
2
238
XM_017025759.1
XP_016881248.1
2
238
XM_017025760.1
XP_016881249.1
2
238
XM_017025761.1
XP_016881250.1
2
238
XM_017025762.1
XP_016881251.1
2
238
XM_017025763.1
XP_016881252.1
2
238
XM_017025764.1
XP_016881253.1
2
238
XM_017025765.1
XP_016881254.1
2
238
XM_017025766.1
XP_016881255.1
2
238
XM_017025767.1
XP_016881256.1
2
238
XM_017025768.1
XP_016881257.1
2
238
XM_017025769.1
XP_016881258.1
2
238
XM_017025770.1
XP_016881259.1
2
238
XM_017025771.1
XP_016881260.1
2
238
XM_017025772.1
XP_016881261.1
2
238
XM_017025773.1
XP_016881262.1
2
238
XM_017025774.1
XP_016881263.1
2
238
XM_017025775.1
XP_016881264.1
2
238
XM_017025776.1
XP_016881265.1
2
238
XM_017025777.1
XP_016881266.1
2
238
GenBank mRNA
AF078830.1
-
1
54
AF078831.1
-
1
54
AF078832.1
-
1
54
AF078833.1
-
1
54
AJ564845.1
-
2
193
AK225889.1
-
2
401
AK225948.1
-
2
197
AK291519.1
-
2
492
AK295401.1
-
2
223
AK300759.1
-
2
272
AK302004.1
-
2
183
BC033242.1
-
2
217
5432943
BC047353.1
-
2
176
DC403953.1
-
2
484
Y10746.1
-
2
193
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
methylation-dependent chromatin silencing
transcription from RNA polymerase II promoter
heart development
aging
response to fluoxetine
neuron differentiation
response to nutrient levels
response to estradiol
response to cocaine
regulation of DNA methylation
negative regulation of transcription, DNA-templated
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
methyl-CpG binding
double-stranded methylated DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0