Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002458.2 | NP_002449.2 | 41, 42 | 16507 | ||
GenBank mRNA | AK303892.1 | - | 8, 9 | 1205 | ||
BM740816.1 | - | 3, 4 | 236 | |||
U78551.1 | - | 4, 5 | 332 | |||
U95031.1 | - | 11, 12 | 1847 |