Search
Search

NME3| Gene Symbol: NME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002513.2 | NP_002504.2 | 2 | 334 | ||
| XM_005255332.4 | XP_005255389.1 | 2 | 365 | |||
| XM_011522503.2 | XP_011520805.1 | 2 | 375 | |||
| XM_011522504.2 | XP_011520806.1 | 2 | 370 | |||
| GenBank mRNA | AK308663.1 | - | 2 | 135 | ||
| BC000250.1 | - | 2 | 334 | 3357930 | ||
| BG911570.1 | - | 2 | 158 | |||
| U29656.1 | - | 2 | 177 | |||