Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006189.1 | NP_006180.1 | 1 | 285 | ||
GenBank mRNA | BC069115.1 | - | 1 | 285 | 7216933 | |
BC069365.1 | - | 1 | 285 | 7262106 |