Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021127.2 | NP_066950.1 | 1 | 230 | ||
GenBank mRNA | AK311943.1 | - | 1 | 195 | ||
BC013120.1 | - | 1 | 153 | 3858958 | ||
BC032663.1 | - | 1 | 185 | 5529754 | ||
BF667112.1 | - | 231 | ||||
D90070.1 | - | 1 | 185 | |||
KU178263.1 | - | 1 | 12 | |||
KU178264.1 | - | 1 | 12 |