Search
Search

PPP1R8| Gene Symbol: PPP1R8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002713.3 | NP_002704.1 | 4 | 802 | ||
| NM_014110.4 | NP_054829.2 | 5 | 621 | |||
| NM_138558.2 | NP_612568.1 | 5 | 903 | |||
| XM_017001594.1 | XP_016857083.1 | 4 | 322 | |||
| GenBank mRNA | AF061958.1 | - | 5 | 549 | ||
| AF061959.1 | - | 5 | 704 | |||
| AF126488.1 | - | 5 | 537 | |||
| AK292077.1 | - | 5 | 549 | |||
| AK297771.1 | - | 4 | 328 | |||
| AK307570.1 | - | 5 | 597 | |||
| AK313814.1 | - | 4 | 308 | |||
| AX775985.1 | - | 5 | 537 | |||
| BC001597.1 | - | 4 | 429 | |||
| BC013360.2 | - | 4 | 479 | 3633676 | ||
| BC093017.1 | - | 5 | 541 | 30531222 | ||
| BI459680.1 | - | 5 | 584 | |||
| DQ223931.1 | - | 2 | 82 | |||
| U14575.1 | - | 4 | 789 | |||