Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278928.1 | NP_001265857.1 | 2, 3 | 276 | ||
NM_017503.4 | NP_059973.4 | 2, 3 | 276 | |||
GenBank mRNA | BC014411.1 | - | 2, 3 | 245 | 4548425 | |
BI118187.1 | - | 2, 3 | 218 | |||
BM789997.1 | - | 2, 3 | 269 | |||
BP244646.1 | - | 2, 3 | 276 | |||
CR456753.1 | - | 2, 3 | 228 | |||
Z35094.1 | - | 2, 3 | 245 |