Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAZ SIRNA Assays ›
          Gene Symbol:
          TAZ
          Gene Name:
          tafazzin
          Gene Aliases:
          BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
          Chromosome Location:
          Chr.X: 154411406 - 154421726 on Build GRCh38
          UniGene ID:
          Hs.409911
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS144201
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAZ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000116.4 NP_000107.1 8 929
          NM_001303465.1 NP_001290394.1 7 941
          NM_181311.3 NP_851828.1 7 839
          NM_181312.3 NP_851829.1 7 887
          NM_181313.3 NP_851830.1 6 797
          NR_024048.2 7 945
          XM_006724836.1 XP_006724899.1 8 960
          XM_006724837.1 XP_006724900.1 6 828
          XM_006724839.1 XP_006724902.1 6 828
          XM_006724841.3 XP_006724904.1 8 681
          XM_006724842.3 XP_006724905.1 7 591
          XM_011531189.1 XP_011529491.1 5 747
          XM_011531191.2 XP_011529493.1 7 1119
          XM_017029761.1 XP_016885250.1 6 774
          XM_017029762.1 XP_016885251.1 7 870
          XM_017029763.1 XP_016885252.1 5 693
          XM_017029764.1 XP_016885253.1 6 458
          XM_017029765.1 XP_016885254.1 6 996
          GenBank mRNA AK130900.1 - 2 1292
          AK291848.1 - 8 797
          AY231461.1 - 7 521
          AY231462.1 - 7 569
          AY231463.1 - 6 479
          AY231464.1 - 8 611
          AY258036.1 - 8 611
          AY258037.1 - 6 479
          AY258038.1 - 7 569
          AY258039.1 - 7 521
          BC005062.1 - 7 902
          BC011515.1 - 5 493 4339983
          BX401320.2 - 6 637
          CA488175.1 - 6 606
          CN478633.1 - 4 460
          DQ884408.1 - 7 615
          KU178348.1 - 5 396
          KU178349.1 - 8 603
          KU178350.1 - 7 561
          KU178351.1 - 6 558
          KU178352.1 - 7 513
          KU178353.1 - 6 471
          X92762.1 - 8 891

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          heart development
          skeletal muscle tissue development
          hemopoiesis
          cardiolipin biosynthetic process
          mitochondrial respiratory chain complex I assembly
          cardiolipin acyl-chain remodeling
          cristae formation
          mitochondrial ATP synthesis coupled electron transport
          cardiac muscle tissue development
          cardiac muscle contraction
          1-acylglycerol-3-phosphate O-acyltransferase activity
          O-acyltransferase activity
          1-acylglycerophosphocholine O-acyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0