Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF7 SIRNA Assays ›
          Gene Symbol:
          TCF7
          Gene Name:
          transcription factor 7 (T-cell specific, HMG-box)
          Gene Aliases:
          TCF-1
          Chromosome Location:
          Chr.5: 134114703 - 134148229 on Build GRCh38
          UniGene ID:
          Hs.573153
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS144220
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCF7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134851.2 NP_001128323.2 7 695
          NM_003202.3 NP_003193.2 8 1139
          NM_201632.3 NP_963963.1 7 695
          NM_201634.3 NP_963965.1 7 695
          NM_213648.3 NP_998813.1 6 929
          NR_033449.1 6 930
          XM_006714678.3 XP_006714741.1 9 1240
          XM_006714679.3 XP_006714742.1 8 1147
          XM_006714680.3 XP_006714743.1 9 1240
          XM_006714682.2 XP_006714745.1 9 1240
          XM_006714684.2 XP_006714747.1 8 1147
          XM_006714685.3 XP_006714748.1 8 785
          XM_006714686.3 XP_006714749.1 7 694
          XM_006714687.3 XP_006714750.1 7 652
          XM_011543604.2 XP_011541906.1 9 1240
          XM_011543606.1 XP_011541908.1 9 1240
          XM_011543607.2 XP_011541909.1 7 6728
          XM_011543608.2 XP_011541910.1 7 2773
          XM_011543609.2 XP_011541911.1 6 641
          XM_011543610.2 XP_011541912.1 7 773
          XM_011543611.2 XP_011541913.1 7 1667
          XM_011543612.2 XP_011541914.1 6 641
          XM_011543613.2 XP_011541915.1 6 680
          XM_017009789.1 XP_016865278.1 8 785
          XM_017009790.1 XP_016865279.1 7 1667
          GenBank mRNA AK057580.1 - 6 930
          AK093530.1 - 6 800
          AK093683.1 - 3 1029
          AK131428.1 - 3 771
          AK310591.1 - 6 641
          AL832816.1 - 8 855
          AL834166.1 - 7 865
          BC048769.1 - 8 1139 6063270
          BC072023.1 - 8 1074 6292932
          X59869.1 - 7 677
          X59870.1 - 7 677
          X59871.1 - 7 677
          Z47361.1 - 6 599
          Z47362.1 - 6 599

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          immune response
          neural tube development
          embryonic genitalia morphogenesis
          T cell receptor V(D)J recombination
          regulation of cell proliferation
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          alpha-beta T cell differentiation
          embryonic digestive tract morphogenesis
          embryonic hindgut morphogenesis
          canonical Wnt signaling pathway
          cellular response to interleukin-4
          beta-catenin-TCF complex assembly
          core promoter binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding
          beta-catenin binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0