Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7 SIRNA Assays ›
Gene Symbol:
TCF7
Gene Name:
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases:
TCF-1
Chromosome Location:
Chr.5: 134114703 - 134148229 on Build GRCh38
UniGene ID:
Hs.573153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS144221
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134851.2
NP_001128323.2
7
735
NM_003202.3
NP_003193.2
8
1179
NM_201632.3
NP_963963.1
7
735
NM_201634.3
NP_963965.1
7
735
NM_213648.3
NP_998813.1
6
969
NR_033449.1
6
970
XM_006714678.3
XP_006714741.1
9
1280
XM_006714679.3
XP_006714742.1
8
1187
XM_006714680.3
XP_006714743.1
9
1280
XM_006714682.2
XP_006714745.1
9
1280
XM_006714684.2
XP_006714747.1
8
1187
XM_006714685.3
XP_006714748.1
8
825
XM_006714686.3
XP_006714749.1
7
734
XM_006714687.3
XP_006714750.1
7
692
XM_011543604.2
XP_011541906.1
9
1280
XM_011543606.1
XP_011541908.1
9
1280
XM_011543607.2
XP_011541909.1
7
6768
XM_011543608.2
XP_011541910.1
7
2813
XM_011543609.2
XP_011541911.1
6
681
XM_011543610.2
XP_011541912.1
7
813
XM_011543611.2
XP_011541913.1
7
1707
XM_011543612.2
XP_011541914.1
6
681
XM_011543613.2
XP_011541915.1
6
720
XM_017009789.1
XP_016865278.1
8
825
XM_017009790.1
XP_016865279.1
7
1707
GenBank mRNA
AK057580.1
-
6
970
AK093530.1
-
6
840
AK093683.1
-
3
1069
AK131428.1
-
3
811
AK310591.1
-
6
681
AL832816.1
-
8
895
AL834166.1
-
7
905
BC048769.1
-
8
1179
6063270
BC072023.1
-
8
1114
6292932
X59869.1
-
7
717
X59870.1
-
7
717
X59871.1
-
7
717
Z47361.1
-
6
639
Z47362.1
-
6
639
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
neural tube development
embryonic genitalia morphogenesis
T cell receptor V(D)J recombination
regulation of cell proliferation
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
alpha-beta T cell differentiation
embryonic digestive tract morphogenesis
embryonic hindgut morphogenesis
canonical Wnt signaling pathway
cellular response to interleukin-4
beta-catenin-TCF complex assembly
core promoter binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-kkgv7:80/100.66.73.110:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0