Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TH SIRNA Assays ›
          Gene Symbol:
          TH
          Gene Name:
          tyrosine hydroxylase
          Gene Aliases:
          DYT14, DYT5b, TYH
          Chromosome Location:
          Chr.11: 2163929 - 2174081 on Build GRCh38
          UniGene ID:
          Hs.435609
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS144270
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000360.3 NP_000351.2 8 944
          NM_199292.2 NP_954986.2 9 1037
          NM_199293.2 NP_954987.2 9 1025
          XM_011520335.2 XP_011518637.1 8 3232
          GenBank mRNA AY144495.1 - 4 219
          BC104967.1 - 8 1016 8143970
          BC143611.1 - 8 1014 9052124
          BC143614.1 - 8 1028 9052127
          BM718799.1 - 5 399
          M17589.1 - 9 1027
          X05290.1 - 8 944
          Y00414.1 - 9 1019

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          synaptic transmission, dopaminergic
          response to amphetamine
          heart morphogenesis
          dopamine biosynthetic process from tyrosine
          fatty acid metabolic process
          sphingolipid metabolic process
          heart development
          visual perception
          sensory perception of sound
          learning
          memory
          mating behavior
          locomotory behavior
          regulation of heart contraction
          response to water deprivation
          response to light stimulus
          response to herbicide
          response to salt stress
          anatomical structure morphogenesis
          organ morphogenesis
          response to zinc ion
          multicellular organism aging
          response to activity
          aminergic neurotransmitter loading into synaptic vesicle
          glycoside metabolic process
          phthalate metabolic process
          cerebral cortex development
          response to nutrient levels
          response to estradiol
          response to lipopolysaccharide
          isoquinoline alkaloid metabolic process
          social behavior
          cellular response to drug
          response to isolation stress
          response to immobilization stress
          neurotransmitter biosynthetic process
          terpene metabolic process
          dopamine biosynthetic process
          epinephrine biosynthetic process
          norepinephrine biosynthetic process
          catecholamine biosynthetic process
          eye photoreceptor cell development
          circadian sleep/wake cycle
          eating behavior
          response to peptide hormone
          pigmentation
          response to ethanol
          response to ether
          response to pyrethroid
          embryonic camera-type eye morphogenesis
          response to corticosterone
          response to electrical stimulus
          phytoalexin metabolic process
          oxidation-reduction process
          cellular response to manganese ion
          cellular response to nicotine
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          tyrosine 3-monooxygenase activity
          protein binding
          ferrous iron binding
          ferric iron binding
          amino acid binding
          oxygen binding
          enzyme binding
          protein domain specific binding
          tetrahydrobiopterin binding
          dopamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0