Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF37A SIRNA Assays ›
          Gene Symbol:
          ZNF37A
          Gene Name:
          zinc finger protein 37A
          Gene Aliases:
          KOX21, ZNF37
          Chromosome Location:
          Chr.10: 38094327 - 38150293 on Build GRCh38
          UniGene ID:
          Hs.292575
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS144501
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF37A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007094.2 NP_001007095.1 5, 6 847
          NM_001178101.1 NP_001171572.1 4, 5 369
          NM_001324245.1 NP_001311174.1 4, 5 298
          NM_001324246.1 NP_001311175.1 5, 6 843
          NM_001324247.1 NP_001311176.1 5, 6 588
          NM_001324248.1 NP_001311177.1 4, 5 553
          NM_001324249.1 NP_001311178.1 4, 5 549
          NM_001324250.1 NP_001311179.1 5, 6 663
          NM_001324251.1 NP_001311180.1 5, 6 667
          NM_001324252.1 NP_001311181.1 5, 6 843
          NM_001324253.1 NP_001311182.1 5, 6 663
          NM_001324254.1 NP_001311183.1 4, 5 373
          NM_001324256.1 NP_001311185.1 5, 6 663
          NM_001324257.1 NP_001311186.1 5, 6 847
          NM_001324258.1 NP_001311187.1 4, 5 369
          NM_001324259.1 NP_001311188.1 5, 6 847
          NM_001324260.1 NP_001311189.1 4, 5 369
          NM_001324261.1 NP_001311190.1 4, 5 369
          NM_001324262.1 NP_001311191.1 5, 6 663
          NM_003421.2 NP_003412.1 4, 5 373
          XM_011519656.2 XP_011517958.1 5, 6 755
          XM_011519657.2 XP_011517959.1 5, 6 577
          XM_011519658.2 XP_011517960.1 4, 5 295
          XM_017016618.1 XP_016872107.1 5, 6 842
          XM_017016619.1 XP_016872108.1 4, 5 378
          XM_017016620.1 XP_016872109.1 5, 6 846
          XM_017016621.1 XP_016872110.1 5, 6 842
          XM_017016622.1 XP_016872111.1 5, 6 842
          XM_017016623.1 XP_016872112.1 5, 6 842
          XM_017016624.1 XP_016872113.1 5, 6 842
          GenBank mRNA AJ492195.1 - 4, 5 337
          AK092012.1 - 4, 5 362
          BC015858.1 - 4, 5 328 4671816
          BX647089.1 - 5, 6 832

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0