Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256171.1 | NP_001243100.1 | 5 | 501 | ||
NM_001256173.1 | NP_001243102.1 | 3 | 284 | |||
NM_003429.4 | NP_003420.2 | 4 | 411 | |||
XM_011528263.2 | XP_011526565.1 | 6 | 572 | |||
XM_011528264.1 | XP_011526566.1 | 2 | 732 | |||
XM_017027234.1 | XP_016882723.1 | 2 | 211 | |||
GenBank mRNA | AK308258.1 | - | 4 | 241 | ||
BC047646.1 | - | 2 | 714 | 4828290 | ||
BP199573.1 | - | 4 | 411 | |||
DB119881.1 | - | 3 | 233 | |||
DC358791.1 | - | 5 | 469 | |||
DN994243.1 | - | 4 | 398 | |||
M61866.1 | - | 4 | 375 | |||
M61868.1 | - | 2 | 131 | |||
U35376.1 | - | 4 | 381 |