Search
Search

HIST1H2BM| Gene Symbol: HIST1H2BM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003521.2 | NP_003512.1 | 1 | 165 | ||
| GenBank mRNA | BC066244.1 | - | 1 | 165 | 7002058 | |
| BC067485.1 | - | 1 | 165 | |||
| BC067486.1 | - | 1 | 165 | 7002060 | ||
| BC067487.1 | - | 1 | 165 | 7002064 | ||
| BC067488.1 | - | 1 | 165 | 7002065 | ||
| BC067489.1 | - | 1 | 165 | 7002066 | ||