Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1G SIRNA Assays ›
Gene Symbol:
CACNA1G
Gene Name:
calcium voltage-gated channel subunit alpha1 G
Gene Aliases:
Ca(V)T.1, Cav3.1, NBR13, SCA42
Chromosome Location:
Chr.17: 50560929 - 50629625 on Build GRCh38
UniGene ID:
Hs.591169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS144927
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256324.1
NP_001243253.1
4
932
NM_001256325.1
NP_001243254.1
4
932
NM_001256326.1
NP_001243255.1
4
932
NM_001256327.1
NP_001243256.1
4
932
NM_001256328.1
NP_001243257.1
4
932
NM_001256329.1
NP_001243258.1
4
932
NM_001256330.1
NP_001243259.1
4
932
NM_001256331.1
NP_001243260.1
4
932
NM_001256332.1
NP_001243261.1
4
932
NM_001256333.1
NP_001243262.1
4
932
NM_001256334.1
NP_001243263.1
4
932
NM_001256359.1
NP_001243288.1
4
932
NM_001256360.1
NP_001243289.1
4
932
NM_001256361.1
NP_001243290.1
4
932
NM_018896.4
NP_061496.2
4
932
NM_198376.2
NP_938190.1
4
932
NM_198377.2
NP_938191.2
4
932
NM_198378.2
NP_938192.1
4
932
NM_198379.2
NP_938193.1
4
932
NM_198380.2
NP_938194.1
4
932
NM_198382.2
NP_938196.1
4
932
NM_198383.2
NP_938197.1
4
932
NM_198384.2
NP_938198.1
4
932
NM_198385.2
NP_938199.1
4
932
NM_198386.2
NP_938200.1
4
932
NM_198387.2
NP_938201.1
4
932
NM_198388.2
NP_938202.1
4
932
NM_198396.2
NP_938406.1
4
932
NR_046054.1
4
932
NR_046055.1
4
932
NR_046056.1
4
932
NR_046057.1
4
932
NR_046058.1
4
932
XM_006722160.3
XP_006722223.1
4
1071
XM_006722161.3
XP_006722224.1
4
1071
GenBank mRNA
AB012043.1
-
4
568
AF124351.1
-
4
912
AF126965.1
-
4
715
AF126966.1
-
4
715
AF134985.1
-
4
540
AF134986.1
-
4
540
AF190860.1
-
4
756
AF227744.1
-
4
540
AF227745.1
-
4
540
AF227746.1
-
4
540
AF227747.1
-
4
540
AF227748.1
-
4
540
AF227749.1
-
4
540
AF227750.1
-
4
540
AF227751.1
-
4
540
BC110995.1
-
4
932
5502027
DQ494449.1
-
4
540
DQ494450.1
-
4
540
DQ494451.1
-
4
540
DQ494452.1
-
4
540
DQ494453.1
-
4
540
DQ494454.1
-
4
540
DQ494455.1
-
4
540
DQ494456.1
-
4
540
DQ494457.1
-
4
540
DQ494458.1
-
4
540
DQ494459.1
-
4
540
DQ494460.1
-
4
540
DQ494461.1
-
4
540
DQ494462.1
-
4
540
DQ494463.1
-
4
540
DQ494464.1
-
4
540
DQ494465.1
-
4
540
DQ494466.1
-
4
540
DQ494467.1
-
4
540
DQ494468.1
-
4
540
DQ494469.1
-
4
540
DQ494470.1
-
4
540
DQ494471.1
-
4
540
DQ494472.1
-
4
540
DQ494473.1
-
4
540
DQ494474.1
-
4
540
DQ494475.1
-
4
540
DQ494476.1
-
4
540
DQ494477.1
-
4
540
DQ494478.1
-
4
540
DQ494479.1
-
4
540
KJ534993.1
-
4
540
KJ535064.1
-
4
523
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
response to nickel cation
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
SA node cell action potential
AV node cell action potential
SA node cell to atrial cardiac muscle cell signalling
AV node cell to bundle of His cell signaling
membrane depolarization during AV node cell action potential
membrane depolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
low voltage-gated calcium channel activity
voltage-gated calcium channel activity involved in AV node cell action potential
voltage-gated calcium channel activity involved SA node cell action potential
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0