Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC28A1 SIRNA Assays ›
Gene Symbol:
SLC28A1
Gene Name:
solute carrier family 28 member 1
Gene Aliases:
CNT1, HCNT1
Chromosome Location:
Chr.15: 84884351 - 84975649 on Build GRCh38
UniGene ID:
Hs.459187
Species:
Human
siRNA ID
HSS145047
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC28A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287761.1
NP_001274690.1
5
442
NM_001287762.1
NP_001274691.1
5
442
NM_001321721.1
NP_001308650.1
6
558
NM_001321722.1
NP_001308651.1
6
558
NM_004213.4
NP_004204.3
6
558
NM_201651.2
NP_964014.1
6
558
XM_011522203.2
XP_011520505.1
6
541
XM_011522204.2
XP_011520506.1
7
1061
XM_011522205.2
XP_011520507.1
6
945
XM_011522206.2
XP_011520508.1
5
744
XM_011522207.2
XP_011520509.1
5
386
XM_011522208.2
XP_011520510.1
4
663
XM_011522209.2
XP_011520511.1
6
541
XM_011522210.2
XP_011520512.1
6
541
XM_011522211.2
XP_011520513.1
5
776
XM_011522212.1
XP_011520514.1
6
541
XM_011522214.2
XP_011520516.1
6
541
XM_011522215.2
XP_011520517.1
6
541
XM_011522216.2
XP_011520518.1
4
297
XM_011522217.1
XP_011520519.1
6
541
XM_011522218.2
XP_011520520.1
6
541
XM_017022723.1
XP_016878212.1
6
541
GenBank mRNA
AF309632.1
-
6
537
AK291997.1
-
6
558
AK296074.1
-
6
557
AK296075.1
-
5
442
AK296119.1
-
5
467
AK298369.1
-
6
558
BC029788.1
-
6
518
BC039898.1
-
5
415
BC126204.1
-
5
448
8991920
BC126206.1
-
5
448
8991922
BC143908.1
-
5
447
U62967.1
-
4
350
U62968.1
-
4
350
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
pyrimidine nucleobase transport
nucleoside transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
purine nucleobase transmembrane transport
nucleoside transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-hf9rh:80/10.239.161.5:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0