Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014760.3 | NP_055575.3 | 4 | 1688 | ||
GenBank mRNA | AK289794.1 | - | 4 | 1409 | ||
BC090935.1 | - | 4 | 1269 | 3064714 | ||
BC101770.1 | - | 4 | 1177 | 8069276 | ||
BC101776.1 | - | 4 | 1178 | 8069282 | ||
D86972.1 | - | 5 | 1505 |