Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HUWE1 SIRNA Assays ›
Gene Symbol:
HUWE1
Gene Name:
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases:
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location:
Chr.X: 53532096 - 53686729 on Build GRCh38
UniGene ID:
Hs.136905
Species:
Human
siRNA ID
HSS145435
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HUWE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031407.6
NP_113584.3
40
5318
XM_005261965.3
XP_005262022.1
39
5385
XM_011530747.1
XP_011529049.1
41
5561
XM_011530751.2
XP_011529053.1
40
5429
XM_017029191.1
XP_016884680.1
39
8313
XM_017029192.1
XP_016884681.1
39
8313
XM_017029193.1
XP_016884682.1
39
8292
XM_017029194.1
XP_016884683.1
39
8313
XM_017029195.1
XP_016884684.1
39
8313
XM_017029196.1
XP_016884685.1
39
8313
XM_017029197.1
XP_016884686.1
39
8313
XM_017029198.1
XP_016884687.1
40
5655
XM_017029199.1
XP_016884688.1
41
5582
XM_017029200.1
XP_016884689.1
40
5452
XM_017029201.1
XP_016884690.1
40
5469
XM_017029202.1
XP_016884691.1
40
5445
XM_017029203.1
XP_016884692.1
40
5451
XM_017029204.1
XP_016884693.1
38
8063
XM_017029205.1
XP_016884694.1
38
8043
XM_017029206.1
XP_016884695.1
39
8313
GenBank mRNA
AB002310.3
-
9
1420
AB071605.1
-
13
2018
AK056671.1
-
23
3544
AK091352.1
-
12
1836
AK294575.1
-
27
3918
AY772009.1
-
38
4963
AY929612.1
-
37
4915
DQ097177.1
-
40
5317
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
base-excision repair
protein monoubiquitination
histone ubiquitination
cell differentiation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
DNA binding
ubiquitin-protein transferase activity
protein binding
ligase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-4vcgr:80/10.239.160.220:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0