Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167947.2 | NP_001161419.1 | 6 | 578 | ||
NM_005834.4 | NP_005825.1 | 5 | 428 | |||
GenBank mRNA | AF034790.1 | - | 5 | 428 | ||
AF077039.1 | - | 4 | 293 | |||
AJ005895.1 | - | 4 | 281 | |||
AK290207.1 | - | 4 | 303 | |||
BC010142.2 | - | 4 | 282 | 4300787 | ||
CB993524.1 | - | 5 | 469 |