Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STUB1 SIRNA Assays ›
          Gene Symbol:
          STUB1
          Gene Name:
          STIP1 homology and U-box containing protein 1
          Gene Aliases:
          CHIP, HSPABP2, NY-CO-7, SCAR16, SDCCAG7, UBOX1
          Chromosome Location:
          Chr.16: 680111 - 682768 on Build GRCh38
          UniGene ID:
          Hs.592081
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS145539
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STUB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001293197.1 NP_001280126.1 3 921
          NM_005861.3 NP_005852.2 3 832
          GenBank mRNA AF039689.1 - 3 473
          AF129085.1 - 3 513
          AF217968.1 - 3 808
          AF432221.1 - 3 528
          BC007545.1 - 3 516 2959735
          BC017178.1 - 3 503 3346811
          BC022788.1 - 3 516 2959735
          BC063617.1 - 3 447 3847168
          CB145452.1 - 3 341

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          DNA repair
          ubiquitin-dependent protein catabolic process
          misfolded or incompletely synthesized protein catabolic process
          protein ubiquitination
          negative regulation of transforming growth factor beta receptor signaling pathway
          ubiquitin-dependent SMAD protein catabolic process
          endoplasmic reticulum unfolded protein response
          positive regulation of protein ubiquitination
          regulation of protein stability
          regulation of glucocorticoid metabolic process
          negative regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          ERAD pathway
          ERBB2 signaling pathway
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of ubiquitin-protein transferase activity
          protein maturation
          protein autoubiquitination
          protein K63-linked ubiquitination
          cellular response to misfolded protein
          positive regulation of chaperone-mediated protein complex assembly
          G-protein coupled receptor binding
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          enzyme binding
          kinase binding
          Hsp70 protein binding
          protein binding, bridging
          TPR domain binding
          ubiquitin protein ligase binding
          ubiquitin-ubiquitin ligase activity
          protein homodimerization activity
          SMAD binding
          misfolded protein binding
          Hsp90 protein binding
          ubiquitin protein ligase activity
          ubiquitin protein ligase activity involved in ERAD pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0