Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144888.1 | NP_001138360.1 | 2 | 185 | ||
NM_006340.2 | NP_006331.1 | 2 | 185 | |||
NM_017450.2 | NP_059344.1 | 2 | 185 | |||
NM_017451.2 | NP_059345.1 | 2 | 185 | |||
XM_005256943.1 | XP_005257000.1 | 3 | 284 | |||
XM_005256944.1 | XP_005257001.1 | 3 | 284 | |||
XM_005256945.1 | XP_005257002.1 | 3 | 284 | |||
XM_005256948.3 | XP_005257005.1 | 2 | 185 | |||
XM_006721635.1 | XP_006721698.1 | 3 | 284 | |||
XM_006721636.1 | XP_006721699.1 | 3 | 284 | |||
XM_006721637.2 | XP_006721700.1 | 2 | 185 | |||
XM_011524193.1 | XP_011522495.1 | 3 | 284 | |||
XM_011524194.1 | XP_011522496.1 | 3 | 284 | |||
GenBank mRNA | AB015019.1 | - | 2 | 170 | ||
AB015020.1 | - | 2 | 170 | |||
AB017119.1 | - | 2 | 170 | |||
AB017120.1 | - | 2 | 170 | |||
AK023688.1 | - | 3 | 280 | |||
AK222670.1 | - | 2 | 181 | |||
AK225612.1 | - | 2 | 182 | |||
AK297644.1 | - | 2 | 177 | |||
BC002495.2 | - | 2 | 160 | |||
BC014020.2 | - | 2 | 172 | 4125501 | ||
BC032559.1 | - | 2 | 183 | 5562784 | ||
CB153324.1 | - | 2 | 210 | |||
U70669.1 | - | 2 | 170 |