Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRG3 SIRNA Assays ›
Gene Symbol:
NRG3
Gene Name:
neuregulin 3
Gene Aliases:
HRG3, pro-NRG3
Chromosome Location:
Chr.10: 81875298 - 82989979 on Build GRCh38
UniGene ID:
Hs.125119
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS145759
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRG3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001010848.3
NP_001010848.2
7
1330
NM_001165972.1
NP_001159444.1
7
1327
NM_001165973.1
NP_001159445.1
8
914
XM_005269444.4
XP_005269501.1
7
1331
XM_011539172.2
XP_011537474.1
7
1333
XM_011539173.2
XP_011537475.1
7
1330
XM_011539175.2
XP_011537477.1
7
1333
XM_011539178.2
XP_011537480.1
4
519
XM_017015573.1
XP_016871062.1
8
1413
XM_017015574.1
XP_016871063.1
8
1411
XM_017015575.1
XP_016871064.1
8
1410
XM_017015576.1
XP_016871065.1
8
1408
XM_017015577.1
XP_016871066.1
8
1413
XM_017015578.1
XP_016871067.1
8
1411
XM_017015579.1
XP_016871068.1
8
1409
XM_017015580.1
XP_016871069.1
8
1408
XM_017015581.1
XP_016871070.1
7
1330
XM_017015582.1
XP_016871071.1
8
683
XM_017015583.1
XP_016871072.1
8
683
GenBank mRNA
AK098823.1
-
6
474
AK122886.1
-
9
1169
AK296563.1
-
7
1152
BC136811.1
-
7
1327
9020801
BC143665.1
-
5
364
DQ001411.1
-
7
1303
DQ338459.1
-
7
582
DQ857894.1
-
8
916
HM068873.1
-
7
1303
HM068874.1
-
5
1073
HM068875.1
-
8
640
HM068876.1
-
7
540
HM068877.1
-
7
540
HM068878.1
-
9
671
HM068879.1
-
7
540
HM068880.1
-
7
615
HM068881.1
-
9
746
HM068882.1
-
8
715
HM068883.1
-
8
715
HM068884.1
-
9
793
HM068885.1
-
5
386
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
activation of transmembrane receptor protein tyrosine kinase activity
pattern specification process
chemorepulsion involved in interneuron migration from the subpallium to the cortex
intracellular signal transduction
animal organ development
mammary placode formation
negative regulation of neuron migration
receptor binding
growth factor activity
transmembrane receptor protein tyrosine kinase activator activity
receptor tyrosine kinase binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0