Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SIPA1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015073.2 | NP_055888.1 | 13 | 4150 | ||
XM_005258671.4 | XP_005258728.1 | 13 | 4591 | |||
XM_011526657.2 | XP_011524959.1 | 13 | 4945 | |||
XM_017026517.1 | XP_016882006.1 | 14 | 5045 | |||
XM_017026518.1 | XP_016882007.1 | 13 | 4547 | |||
GenBank mRNA | AB011117.2 | - | 11 | 2396 | ||
AY168880.1 | - | 11 | 3896 | |||
BC150620.1 | - | 13 | 4129 | 9056990 |