Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009880.1 | NP_001009880.1 | 7 | 839 | ||
NM_015264.1 | NP_056079.1 | 7 | 911 | |||
XM_011530017.2 | XP_011528319.1 | 7 | 874 | |||
XM_017028672.1 | XP_016884161.1 | 7 | 883 | |||
GenBank mRNA | AB023147.1 | - | 6 | 607 | ||
AK025608.1 | - | 5 | 428 | |||
AK094937.1 | - | 8 | 922 | |||
AK124648.1 | - | 7 | 919 | |||
AK297058.1 | - | 7 | 891 | |||
AK309015.1 | - | 6 | 963 | |||
AK315883.1 | - | 7 | 823 | |||
BC016882.1 | - | 2 | 169 | |||
BC038414.1 | - | 6 | 986 | |||
BC053595.1 | - | 7 | 833 | 5730020 | ||
CR456402.1 | - | 7 | 776 | |||
KC154032.1 | - | 7 | 839 |