Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 3 | 374 | ||
NM_015303.3 | NP_056118.2 | 3 | 374 | |||
XM_005247251.3 | XP_005247308.1 | 3 | 642 | |||
XM_005247253.4 | XP_005247310.1 | 3 | 380 | |||
XM_017006040.1 | XP_016861529.1 | 2 | 68 | |||
XM_017006041.1 | XP_016861530.1 | 3 | 380 | |||
GenBank mRNA | AK056661.1 | - | 3 | 346 | ||
AK292423.1 | - | 3 | 346 | |||
BC140768.1 | - | 3 | 310 | 9021636 | ||
BX647915.1 | - | 3 | 336 | |||
DA218768.1 | - | 3 | 381 | |||
DA340096.1 | - | 3 | 331 |