Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002880.1 | NP_001002880.2 | 6 | 596 | ||
NM_015373.3 | NP_056188.1 | 5 | 456 | |||
GenBank mRNA | AB111855.1 | - | 4 | 334 | ||
AF331041.1 | - | 5 | 392 | |||
AL050345.1 | - | 5 | 435 | |||
AL136686.1 | - | 5 | 439 | |||
BC016139.1 | - | 5 | 434 | 3922707 | ||
CR456410.1 | - | 4 | 348 |