Search
Search

IL37| Gene Symbol: IL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014439.3 | NP_055254.2 | 5 | 466 | ||
| NM_173202.1 | NP_775294.1 | 4 | 403 | |||
| NM_173203.1 | NP_775295.1 | 3 | 283 | |||
| NM_173204.1 | NP_775296.1 | 4 | 346 | |||
| NM_173205.1 | NP_775297.1 | 4 | 346 | |||
| XM_011510963.2 | XP_011509265.1 | 6 | 2467 | |||
| XM_011510964.2 | XP_011509266.1 | 5 | 2714 | |||
| GenBank mRNA | AB451333.1 | - | 4 | 361 | ||
| AB451478.1 | - | 4 | 361 | |||
| AF167368.1 | - | 5 | 466 | |||
| AF200496.1 | - | 5 | 472 | |||
| AF201832.1 | - | 4 | 346 | |||
| AF251118.1 | - | 5 | 496 | |||
| AF251119.1 | - | 5 | 424 | |||
| AF251120.1 | - | 4 | 304 | |||
| AY071840.1 | - | 4 | 361 | |||
| AY071841.1 | - | 3 | 241 | |||
| BC020637.1 | - | 5 | 427 | 4731213 | ||