Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC35C2 SIRNA Assays ›
Gene Symbol:
SLC35C2
Gene Name:
solute carrier family 35 member C2
Gene Aliases:
BA394O2.1, C20orf5, CGI-15, OVCOV1
Chromosome Location:
Chr.20: 46349528 - 46364461 on Build GRCh38
UniGene ID:
Hs.593344
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS147227
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC35C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281457.1
NP_001268386.1
2
450
NM_001281458.1
NP_001268387.1
3
318
NM_001281459.1
NP_001268388.1
2
450
NM_001281460.1
NP_001268389.1
3
782
NM_015945.11
NP_057029.8
2
450
NM_173073.3
NP_775096.1
2
450
NM_173179.3
NP_775271.1
2
645
XM_011528831.1
XP_011527133.1
2
628
XM_011528832.1
XP_011527134.1
2
922
XM_011528833.1
XP_011527135.1
2
446
XM_011528834.1
XP_011527136.1
2
450
XM_011528835.1
XP_011527137.1
2
450
XM_011528836.1
XP_011527138.1
2
602
XM_011528837.1
XP_011527139.1
2
436
XM_011528838.1
XP_011527140.1
1
557
XM_017027856.1
XP_016883345.1
2
1345
XM_017027857.1
XP_016883346.1
2
873
XM_017027858.1
XP_016883347.1
2
783
XM_017027859.1
XP_016883348.1
2
1150
XM_017027860.1
XP_016883349.1
3
590
XM_017027861.1
XP_016883350.1
2
612
XM_017027862.1
XP_016883351.1
2
450
GenBank mRNA
AF132949.1
-
2
387
AF455052.1
-
2
405
AK222928.1
-
2
390
AK300207.1
-
2
404
AK300798.1
-
2
382
AK301022.1
-
3
260
AK315726.1
-
2
612
BC014191.2
-
2
394
4761663
BC021138.1
-
2
588
4870715
BC025277.1
-
2
646
4865888
BI831689.1
-
3
394
BM846744.1
-
2
416
DA820126.1
-
2
450
DB023133.1
-
3
533
DB246755.1
-
2
405
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
negative regulation of gene expression
UDP-glucose transport
protein O-linked fucosylation
positive regulation of Notch signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline