Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321025.1 | NP_001307954.1 | 2 | 524 | ||
NM_001321026.1 | NP_001307955.1 | 2 | 582 | |||
NM_016074.4 | NP_057158.1 | 2 | 357 | |||
XM_006711348.3 | XP_006711411.1 | 2 | 390 | |||
GenBank mRNA | AF151901.1 | - | 2 | 292 | ||
AK313014.1 | - | 1 | 673 | |||
BC002771.2 | - | 2 | 317 | |||
BC063405.1 | - | 2 | 317 | 5190565 | ||
BF970430.1 | - | 538 | ||||
CA389141.1 | - | 2 | 337 | |||
CB992246.1 | - | 2 | 500 |