Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015982.3 | NP_057066.2 | 4 | 489 | ||
XM_017024713.1 | XP_016880202.1 | 5 | 579 | |||
GenBank mRNA | AB208818.1 | - | 1 | 3470 | ||
AF096834.1 | - | 4 | 487 | |||
AK092747.1 | - | 3 | 1106 | |||
AK315035.1 | - | 4 | 460 | |||
BC033800.1 | - | 4 | 452 | 5171696 | ||
BC047760.1 | - | 4 | 453 | 5742121 |