Search
Search

AHSP| Gene Symbol: AHSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318221.1 | NP_001305150.1 | 2, 3 | 177 | ||
| NM_001318222.1 | NP_001305151.1 | 2, 3 | 108 | |||
| NM_016633.3 | NP_057717.1 | 2, 3 | 181 | |||
| GenBank mRNA | AF112995.1 | - | 2, 3 | 212 | ||
| AF147435.1 | - | 2, 3 | 140 | |||
| AF208865.1 | - | 2, 3 | 137 | |||
| AF364517.1 | - | 2, 3 | 137 | |||
| BC035842.1 | - | 2, 3 | 122 | 5748040 | ||
| BG940214.1 | - | 2, 3 | 136 | |||
| BG943964.1 | - | 2, 3 | 130 | |||
| H94920.1 | - | 2, 3 | 170 | |||
| R29304.1 | - | 2, 3 | 107 | |||