Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC7 SIRNA Assays ›
          Gene Symbol:
          HDAC7
          Gene Name:
          histone deacetylase 7
          Gene Aliases:
          HD7, HD7A, HDAC7A
          Chromosome Location:
          Chr.12: 47782710 - 47820612 on Build GRCh38
          UniGene ID:
          Hs.200063
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS147499
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098416.3 NP_001091886.1 11, 12 1543
          NM_001308090.1 NP_001295019.1 11, 12 1603
          NM_015401.4 NP_056216.2 12, 13 1654
          XM_011538478.1 XP_011536780.1 12, 13 1606
          XM_011538479.1 XP_011536781.1 13, 14 2048
          XM_011538480.1 XP_011536782.1 11, 12 1495
          XM_011538481.1 XP_011536783.1 12, 13 1499
          XM_011538482.1 XP_011536784.1 12, 13 1583
          XM_011538483.1 XP_011536785.1 12, 13 1606
          XM_017019455.1 XP_016874944.1 11, 12 1555
          XM_017019456.1 XP_016874945.1 12, 13 1606
          GenBank mRNA AF239243.1 - 9, 10 1437
          AK001190.1 - 4, 5 576
          AK024469.1 - 7, 8 969
          AK299292.1 - 12, 13 1569
          AK301545.1 - 12, 13 1517
          AK303481.1 - 11, 12 1509
          AL117455.1 - 8, 9 1121
          AY302468.1 - 12, 13 1459
          AY321367.1 - 11, 12 1428
          BC006453.1 - 1, 2 78
          BC020505.2 - 4, 5 621
          BC064840.1 - 11, 12 1437 6179239
          BT009771.1 - 2, 3 328

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculogenesis
          transcription, DNA-templated
          cell-cell junction assembly
          chromatin modification
          protein sumoylation
          neuron differentiation
          negative regulation of interleukin-2 production
          negative regulation of osteoblast differentiation
          neuron apoptotic process
          histone H3 deacetylation
          cellular response to mechanical stimulus
          positive regulation of cell migration involved in sprouting angiogenesis
          negative regulation of neuron death
          negative regulation of NIK/NF-kappaB signaling
          chromatin binding
          transcription corepressor activity
          protein kinase C binding
          protein binding
          SUMO transferase activity
          protein kinase binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          activating transcription factor binding
          metal ion binding
          repressing transcription factor binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0