Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS147792
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
4
437
NM_001195249.1
NP_001182178.1
4
606
NM_001195250.1
NP_001182179.1
3
275
NM_001195251.1
NP_001182180.1
5
543
NM_001195254.1
NP_001182183.1
3
297
NM_175069.2
NP_778239.1
4
437
NM_175073.2
NP_778243.1
5
543
NR_036576.1
4
432
NR_036577.1
4
322
NR_036578.1
4
454
NR_036579.1
4
601
XM_006716791.3
XP_006716854.1
4
445
XM_006716792.2
XP_006716855.1
4
463
XM_011517938.1
XP_011516240.1
3
421
XM_011517939.2
XP_011516241.1
3
376
XM_017014831.1
XP_016870320.1
4
437
XM_017014832.1
XP_016870321.1
4
437
XM_017014833.1
XP_016870322.1
4
437
XM_017014834.1
XP_016870323.1
4
437
XM_017014835.1
XP_016870324.1
4
437
XM_017014836.1
XP_016870325.1
4
437
XM_017014837.1
XP_016870326.1
3
526
XM_017014838.1
XP_016870327.1
4
419
GenBank mRNA
AB603741.1
-
3
333
AJ565850.1
-
4
295
AJ565851.1
-
4
300
AJ565852.1
-
4
410
AJ565853.1
-
3
413
AJ565854.1
-
3
418
AJ565855.1
-
2
256
AJ575566.1
-
3
298
AK000164.1
-
3
440
AK055672.1
-
4
424
AK131046.1
-
4
389
AY040777.1
-
4
412
AY208829.1
-
4
412
AY208830.1
-
3
275
AY208832.1
-
3
250
AY208833.1
-
3
250
AY208834.1
-
4
408
AY208835.1
-
4
392
AY208836.1
-
4
552
AY208837.1
-
5
509
AY208838.1
-
3
368
AY208839.1
-
4
546
AY208840.1
-
4
409
AY208841.1
-
3
246
AY208842.1
-
1
46
AY302067.1
-
3
555
AY302068.1
-
3
550
AY302069.1
-
3
440
AY302070.1
-
3
435
AY302071.1
-
3
555
AY302072.1
-
3
550
AY302073.1
-
3
440
AY302074.1
-
3
435
BC001628.1
-
3
260
BC032650.1
-
4
478
5505120
BC068107.1
-
5
518
BC104881.1
-
4
451
8143884
BQ957536.1
-
5
526
BX538161.1
-
5
522
DA664623.1
-
3
464
DB042208.1
-
3
297
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0