Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190266.1 | NP_001177195.1 | 17 | 1919 | ||
NM_001190267.1 | NP_001177196.1 | 17 | 1917 | |||
NM_017974.3 | NP_060444.3 | 16 | 1864 | |||
NM_030803.6 | NP_110430.5 | 17 | 1921 | |||
NM_198890.2 | NP_942593.2 | 13 | 1432 | |||
XM_005246082.2 | XP_005246139.1 | 18 | 1909 | |||
XM_005246084.2 | XP_005246141.1 | 15 | 1448 | |||
XM_005246086.2 | XP_005246143.1 | 14 | 1397 | |||
XM_006712608.2 | XP_006712671.1 | 17 | 1720 | |||
GenBank mRNA | AK001690.1 | - | 16 | 1709 | ||
AK024453.1 | - | 15 | 2732 | |||
AK026330.1 | - | 4 | 399 | |||
AK027854.1 | - | 15 | 1446 | |||
AK074434.1 | - | 11 | 891 | |||
AK123876.1 | - | 14 | 1283 | |||
AK308400.1 | - | 16 | 1760 | |||
AL834526.1 | - | 3 | 129 | |||
AY358182.1 | - | 16 | 2083 | |||
AY398617.1 | - | 17 | 1810 | |||
BC071846.1 | - | 13 | 1270 | |||
BC117337.1 | - | 17 | 1583 | |||
BC143900.1 | - | 16 | 1526 | |||
EF079889.1 | - | 16 | 1607 | |||
EF079890.1 | - | 13 | 1175 | |||
EF560734.1 | - | 16 | 1734 | |||
JQ924062.1 | - | 16 | 1607 |