Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035223.3 | NP_001030300.3 | 18, 19 | 2084 | ||
NM_001161616.2 | NP_001155088.2 | 18, 19 | 2102 | |||
GenBank mRNA | AK096811.1 | - | 12, 13 | 1689 | ||
BC014426.1 | - | 18, 19 | 2070 | |||
BC101756.1 | - | 18, 19 | 2070 | 8069262 | ||
BC111958.1 | - | 18, 19 | 2070 | 8327426 | ||
BC143530.1 | - | 18, 19 | 2088 | 9052039 | ||
BM992649.1 | - | 1, 2 | 474 |