Search
Search

USP35| Gene Symbol: USP35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020798.2 | NP_065849.1 | 11 | 3385 | ||
| XM_011545187.2 | XP_011543489.1 | 11 | 2528 | |||
| XM_011545188.2 | XP_011543490.1 | 9 | 2311 | |||
| XM_017018052.1 | XP_016873541.1 | 11 | 3382 | |||
| XM_017018053.1 | XP_016873542.1 | 10 | 2377 | |||
| GenBank mRNA | AB037793.1 | - | 8 | 2848 | ||
| AJ586137.1 | - | 10 | 3026 | |||
| AK056231.1 | - | 6 | 2178 | |||
| AK301048.1 | - | 3 | 1776 | |||
| BC131489.1 | - | 9 | 2352 | 40038029 | ||