Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020862.1 | NP_065913.1 | 1 | 1304 | ||
XM_005259102.4 | XP_005259159.1 | 4 | 1504 | |||
XM_017027033.1 | XP_016882522.1 | 4 | 1619 | |||
GenBank mRNA | AB040917.1 | - | 1 | 1093 | ||
BC014678.1 | - | 400 | ||||
BC025310.1 | - | 1 | 617 |