Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260476.2 | XP_005260533.1 | 5 | 3446 | ||
XM_006723841.2 | XP_006723904.1 | 3 | 3092 | |||
XM_011528952.2 | XP_011527254.1 | 5 | 3507 | |||
XM_011528953.2 | XP_011527255.1 | 4 | 3387 | |||
GenBank mRNA | AJ297792.1 | - | 5 | 3454 | ||
AK025412.1 | - | 1 | 1179 | |||
AK027153.1 | - | 1 | 1260 | |||
AL122050.1 | - | 1 | 797 | |||
AL137597.1 | - | 1 | 2274 | |||
AY422170.1 | - | 5 | 3463 | |||
BC035639.1 | - | 5 | 3353 | 5547220 |