Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 14 | 1833 | ||
XM_005273602.3 | XP_005273659.1 | 14 | 1833 | |||
XM_006716381.2 | XP_006716444.1 | 14 | 1833 | |||
XM_006716385.3 | XP_006716448.1 | 15 | 3328 | |||
XM_017013732.1 | XP_016869221.1 | 14 | 1830 | |||
XM_017013733.1 | XP_016869222.1 | 15 | 1950 | |||
XM_017013734.1 | XP_016869223.1 | 14 | 1814 | |||
GenBank mRNA | AF495722.1 | - | 9 | 1240 | ||
AK025454.1 | - | 3 | 243 | |||
AK055191.1 | - | 8 | 892 | |||
AK095924.1 | - | 2 | 123 | |||
AK315634.1 | - | 2 | 99 | |||
BC012865.1 | - | 5 | 646 | |||
BC013350.2 | - | 4 | 324 | |||
BC052237.1 | - | 14 | 1830 | |||
BC112147.1 | - | 8 | 922 |