Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323680.1 | NP_001310609.1 | 4 | 848 | ||
NM_001323681.1 | NP_001310610.1 | 4 | 839 | |||
NM_001323682.1 | NP_001310611.1 | 5 | 905 | |||
NM_001323683.1 | NP_001310612.1 | 5 | 899 | |||
NM_001323684.1 | NP_001310613.1 | 5 | 927 | |||
NM_001323685.1 | NP_001310614.1 | 4 | 568 | |||
NM_024958.2 | NP_079234.1 | 4 | 826 | |||
NR_136649.1 | 3 | 631 | ||||
XM_006723630.3 | XP_006723693.1 | 4 | 1249 | |||
XM_011529360.1 | XP_011527662.1 | 4 | 532 | |||
XM_011529362.2 | XP_011527664.1 | 5 | 539 | |||
XM_011529363.2 | XP_011527665.1 | 4 | 517 | |||
XM_017028073.1 | XP_016883562.1 | 4 | 471 | |||
XM_017028074.1 | XP_016883563.1 | 4 | 525 | |||
XM_017028075.1 | XP_016883564.1 | 4 | 697 | |||
GenBank mRNA | AK026982.1 | - | 4 | 439 | ||
AK290527.1 | - | 4 | 803 | |||
AL136915.1 | - | 3 | 421 | |||
BC001963.1 | - | 3 | 406 | 3454427 | ||
BE792891.1 | - | 2 | 190 | |||
BP202782.1 | - | 5 | 520 | |||
CD102518.1 | - | 4 | 457 | |||
CR533441.1 | - | 2 | 288 | |||
DA479487.1 | - | 4 | 476 |