Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM103A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031452.3 | NP_113640.1 | 4 | 566 | ||
GenBank mRNA | AK027095.1 | - | 4 | 527 | ||
AL049237.1 | - | 1 | 131 | |||
BC003627.2 | - | 1 | 526 | 2989772 | ||
BC027181.1 | - | 1 | 518 | 3924570 | ||
BC112329.1 | - | 1 | 535 | 5578103 |