Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032023.3 | NP_114412.2 | 2, 3 | 206 | ||
XM_005271833.4 | XP_005271890.1 | 2, 3 | 175 | |||
XM_005271835.4 | XP_005271892.1 | 2, 3 | 175 | |||
XM_006718013.3 | XP_006718076.1 | 2, 3 | 175 | |||
XM_011540255.2 | XP_011538557.1 | 2, 3 | 224 | |||
XM_017016756.1 | XP_016872245.1 | 2, 3 | 175 | |||
GenBank mRNA | AF260335.1 | - | 2, 3 | 170 | ||
AY216713.1 | - | 2, 3 | 206 | |||
AY216716.1 | - | 2, 3 | 206 | |||
BC032593.1 | - | 2, 3 | 188 | 5527647 | ||
BI601296.1 | - | 2, 3 | 209 |