Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032871.3 | NP_116260.2 | 10 | 1306 | ||
NM_152222.1 | NP_689408.1 | 10 | 1244 | |||
XM_005274356.2 | XP_005274413.1 | 5 | 1407 | |||
XM_006718714.2 | XP_006718777.1 | 8 | 871 | |||
XM_011545305.1 | XP_011543607.1 | 9 | 1129 | |||
GenBank mRNA | AF319553.1 | - | 10 | 1306 | ||
AK027899.1 | - | 10 | 1244 | |||
AK057036.1 | - | 4 | 1048 | |||
AK074128.1 | - | 2 | 2979 | |||
AK310103.1 | - | 7 | 1112 | |||
BC017279.2 | - | 10 | 1169 | 4873951 | ||
BC051810.1 | - | 10 | 1196 | 5216988 |