Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080593.2 | NP_542160.1 | 2 | 807 | ||
GenBank mRNA | AJ223352.1 | - | 2 | 762 | ||
BC000893.1 | - | 2 | 789 | 3463930 | ||
BC051872.1 | - | 2 | 787 | 6526835 | ||
BC064959.1 | - | 2 | 787 | 6093977 | ||
BC108737.1 | - | 2 | 787 | 4707078 |