Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271082.1 | NP_001258011.1 | 5, 6 | 417 | ||
NM_033120.3 | NP_149111.1 | 5, 6 | 417 | |||
XM_005248382.3 | XP_005248439.1 | 5, 6 | 534 | |||
XM_005248383.3 | XP_005248440.1 | 5, 6 | 534 | |||
XM_011514150.2 | XP_011512452.1 | 5, 6 | 534 | |||
GenBank mRNA | AB062887.1 | - | 5, 6 | 326 | ||
AF358137.1 | - | 5, 6 | 338 | |||
BC012176.1 | - | 5, 6 | 417 | 4649935 |