Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCDC189 SIRNA Assays ›
          Gene Symbol:
          CCDC189 PHKG2
          Gene Name:
          coiled-coil domain containing 189
          phosphorylase kinase catalytic subunit gamma 2
          Gene Aliases:
          C16orf93
          GSD9C
          Chromosome Location:
          Chr.16: 30757226 - 30762277 on Build GRCh38
          Chromosome Location:
          Chr.16: 30748299 - 30761176 on Build GRCh38
          UniGene ID:
          Hs.724010
          Hs.65735
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS150526
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCDC189
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014979.2 NP_001014979.2 9 1457
          XM_011545976.1 XP_011544278.1 8 1400
          XM_011545978.1 XP_011544280.1 6 1245
          XM_017023852.1 XP_016879341.1 8 1685
          XM_017023853.1 XP_016879342.1 7 1622
          XM_017023854.1 XP_016879343.1 7 1595
          XM_017023855.1 XP_016879344.1 9 1589
          XM_017023856.1 XP_016879345.1 8 1206
          XM_017023857.1 XP_016879346.1 7 1676
          XM_017023858.1 XP_016879347.1 6 1525
          XM_017023859.1 XP_016879348.1 7 1401
          XM_017023860.1 XP_016879349.1 5 860
          XM_017023861.1 XP_016879350.1 8 1471
          XM_017023862.1 XP_016879351.1 7 2003
          GenBank mRNA AL122109.2 - 6 2039
          BC042548.1 - 6 1672
          BC092491.1 - 8 1749 4837696
          BC128128.1 - 8 1113
          BC128129.1 - 9 961 40112368
          Gene Symbol: PHKG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000294.2 NP_000285.1 10 1834
          GenBank mRNA AB209055.1 - 5 1223

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          glycogen biosynthetic process
          glycogen catabolic process
          generation of precursor metabolites and energy
          protein phosphorylation
          positive regulation of glycogen catabolic process
          protein serine/threonine kinase activity
          phosphorylase kinase activity
          calmodulin binding
          ATP binding
          enzyme binding
          tau-protein kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0