Search
Search

TMEM261| Gene Symbol: TMEM261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033428.2 | NP_219500.1 | 1, 2 | 337 | ||
| GenBank mRNA | AI284226.1 | - | 1, 2 | 306 | ||
| AI310403.1 | - | 1, 2 | 304 | |||
| AK292632.1 | - | 1, 2 | 309 | |||
| AL137489.1 | - | 1, 2 | 141 | |||
| BI550091.1 | - | 1, 2 | 337 | |||
| DA887295.1 | - | 1, 2 | 337 | |||